The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_146039.3 | NP_666151.3 | 4 - 5 | 745 | 132 |
GenBank mRNA | AK046399.1 | - | 4 - 5 | 707 | 132 |
AK052482.1 | - | 4 - 5 | 709 | 132 | |
AK144138.1 | - | 4 - 5 | 745 | 132 | |
BC138916.1 | - | 4 - 5 | 678 | 132 |