The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172881.3 | NP_766469.1 | 5 - 6 | 1344 | 119 |
GenBank mRNA | AK083294.1 | - | 5 - 6 | 1344 | 119 |
AK149501.1 | - | 5 - 6 | 1331 | 119 | |
BC113789.1 | - | 5 - 6 | 1334 | 119 |