The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009236.2 | NP_033262.2 | 1 - 2 | 455 | 73 |
GenBank mRNA | AK144281.1 | - | 1 - 2 | 456 | 73 |
BC006612.1 | - | 1 - 2 | 455 | 73 | |
L29086.1 | - | 1 - 2 | 108 | 73 | |
L35032.1 | - | 1 - 2 | 424 | 73 |