The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001103165.1 | NP_001096635.1 | 9 - 10 | 913 | 140 |
NM_011042.2 | NP_035172.2 | 9 - 10 | 913 | 140 | |
GenBank mRNA | AK012297.1 | - | 9 - 10 | 760 | 140 |
AK169268.1 | - | 9 - 10 | 1007 | 140 | |
EU604548.1 | - | 8 - 9 | 668 | 140 | |
L19661.1 | - | 9 - 10 | 800 | 140 |