Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Thrap3
- Entrez Gene ID
-
230753
- Gene Name
- thyroid hormone receptor associated protein 3
- Gene Aliases
-
9330151F09Rik, B230333E16Rik, Trap150
- Chromosome Location
-
Chr.4: 126164082 - 126202710 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK034044.1 |
- |
6 - 7 |
2056 |
157 |
View Full Product Details