The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_139151.4 | NP_631890.3 | 5 - 6 | 742 | 127 |
GenBank mRNA | AB041554.1 | - | 3 - 4 | 1321 | 127 |
AK028080.1 | - | 5 - 6 | 711 | 127 | |
AK043812.1 | - | 5 - 6 | 710 | 127 |