Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Epn2
Entrez Gene ID
13855
Gene Name
epsin 2
Gene Aliases
9530051D10Rik, AA536924, Ibp2
Chromosome Location
Chr.11: 61517249 - 61579881 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001252188.1 NP_001239117.1 9 - 10 1776 140
NM_001252189.1 NP_001239118.1 7 - 8 1532 140
NM_010148.3 NP_034278.1 8 - 9 1568 140
GenBank mRNA AF057286.1 - 6 - 7 1023 140
AK036331.1 - 7 - 8 1506 140
AK139234.1 - 9 - 10 1764 140
AK139793.1 - 9 - 10 1699 140
AK160782.1 - 7 - 8 1533 140
AK173093.1 - 9 - 10 1752 140
BC039138.1 - 8 - 9 1532 140

View Full Product Details