The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009274.2 | NP_033300.2 | 1 - 2 | 242 | 87 |
GenBank mRNA | AK136861.1 | - | 1 - 2 | 207 | 87 |
AK161384.1 | - | 2 - 3 | 301 | 87 | |
AK169580.1 | - | 1 - 2 | 215 | 87 | |
BB860938.1 | - | 1 - 2 | 250 | 87 | |
BC020178.1 | - | 1 - 2 | 139 | 87 |