Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Trps1
Entrez Gene ID
83925
Gene Name
trichorhinophalangeal syndrome I (human)
Gene Aliases
AI115454, AI447310, D15Ertd586e
Chromosome Location
Chr.15: 50654757 - 50890747 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC049857.1 - 3 - 4 352 59

View Full Product Details