Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Vps26b
Entrez Gene ID
69091
Gene Name
vacuolar protein sorting 26 homolog B (yeast)
Gene Aliases
1810012I05Rik, 2310075A12Rik, AI848392
Chromosome Location
Chr.9: 27004502 - 27030094 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178027.4 NP_821170.1 1 - 2 695 78
GenBank mRNA AK031589.1 - 1 - 2 698 78
AK035321.1 - 1 - 2 648 78
AK150929.1 - 1 - 2 616 78
AK155578.1 - 1 - 2 656 78
AK165539.1 - 1 - 2 634 78
AK171154.1 - 1 - 2 654 78
AK185841.1 - 1 - 2 149 78
AK192911.1 - 1 - 2 151 78
AK194762.1 - 1 - 2 149 78
BC049180.1 - 1 - 2 658 78
BC052721.1 - 1 - 2 619 78
BC062972.1 - 1 - 2 619 78

View Full Product Details