The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_181596.4 | NP_853627.2 | 4 - 4 | 532 | 74 |
GenBank mRNA | AB086819.1 | - | 3 - 3 | 453 | 74 |
AJ514933.1 | - | 3 - 3 | 456 | 74 | |
AJ536019.1 | - | 4 - 4 | 495 | 74 | |
AK158401.1 | - | 4 - 4 | 496 | 74 | |
BX631439.1 | - | 1 - 1 | 74 |