The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172625.2 | NP_766213.1 | 2 - 3 | 284 | 77 |
GenBank mRNA | AK077290.1 | - | 2 - 3 | 296 | 77 |
AK083718.1 | - | 2 - 3 | 285 | 77 | |
BC060961.1 | - | 2 - 3 | 302 | 77 |