Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Brcc3
Entrez Gene ID
210766
Gene Name
BRCA1/BRCA2-containing complex, subunit 3
Gene Aliases
C6.1A
Chromosome Location
Chr.X: 75416628 - 75455702 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001166457.1 NP_001159929.1 3 - 4 291 73
NM_001166459.1 NP_001159931.1 3 - 4 291 73
NM_145956.4 NP_666068.1 3 - 4 291 73
GenBank mRNA AK011876.1 - 3 - 4 208 73
AK028483.1 - 1 - 2 1773 73
AK030736.1 - 3 - 4 221 73
AK038919.1 - 3 - 4 209 73
AK149844.1 - 3 - 4 223 73
BC021313.1 - 3 - 4 238 73
BC048179.1 - 3 - 4 205 73
S68022.1 - 3 - 4 233 73

View Full Product Details