The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173376.3 | NP_775552.1 | 6 - 6 | 1054 | 116 |
GenBank mRNA | AK076115.1 | - | 6 - 6 | 1055 | 116 |
AK168018.1 | - | 6 - 6 | 969 | 116 | |
BC026976.1 | - | 6 - 6 | 979 | 116 |