The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007592.3 | NP_031618.2 | 1 - 2 | 196 | 134 |
GenBank mRNA | AK004896.1 | - | 1 - 2 | 197 | 134 |
AK004923.1 | - | 1 - 2 | 197 | 134 | |
BC010773.1 | - | 1 - 2 | 172 | 134 |