Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Scn7a
Entrez Gene ID
20272
Gene Name
sodium channel, voltage-gated, type VII, alpha
Gene Aliases
1110034K09Rik, AI642000, NaG, Nav2, Nav2.3, Nax, Scn6a
Chromosome Location
Chr.2: 66673424 - 66785091 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009135.2 NP_033161.2 23 - 24 3926 151
GenBank mRNA BC145273.1 - 23 - 24 3782 151
L36179.1 - 23 - 24 3962 151

View Full Product Details