The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001136070.1 | NP_001129542.1 | 8 - 9 | 1076 | 82 |
NM_010709.3 | NP_034839.2 | 8 - 9 | 1076 | 82 | |
GenBank mRNA | AK034115.1 | - | 8 - 9 | 1048 | 82 |
AK036154.1 | - | 8 - 9 | 1072 | 82 | |
AK046539.1 | - | 8 - 9 | 1088 | 82 | |
AK047005.1 | - | 8 - 9 | 1071 | 82 | |
AK082748.1 | - | 8 - 9 | 1091 | 82 | |
AK083638.1 | - | 8 - 9 | 1036 | 82 | |
AK139052.1 | - | 8 - 9 | 1076 | 82 | |
AK161269.1 | - | 8 - 9 | 1059 | 82 | |
AK166085.1 | - | 8 - 9 | 1089 | 82 | |
AK167980.1 | - | 8 - 9 | 1072 | 82 | |
AK170252.1 | - | 8 - 9 | 1103 | 82 | |
AK170448.1 | - | 8 - 9 | 1088 | 82 | |
AK171115.1 | - | 8 - 9 | 1054 | 82 | |
BC023709.1 | - | 8 - 9 | 1029 | 82 | |
BC024501.1 | - | 3 - 4 | 390 | 82 | |
BC025036.1 | - | 8 - 9 | 1032 | 82 | |
U58337.1 | - | 8 - 9 | 961 | 82 |