Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tulp1
Entrez Gene ID
22157
Gene Name
tubby like protein 1
Gene Aliases
Tulp1l
Chromosome Location
Chr.17: 28351515 - 28365211 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021478.1 NP_067453.1 2 - 3 122 137
GenBank mRNA AF085681.1 - 2 - 3 170 137
AF105711.1 - 2 - 3 170 137
BC014820.1 - 2 - 3 127 137
BC029222.1 - 2 - 3 161 137

View Full Product Details