The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001141975.1 | NP_001135447.1 | 4 - 5 | 521 | 118 |
NM_001141976.1 | NP_001135448.1 | 4 - 5 | 537 | 118 | |
NM_001141977.1 | NP_001135449.1 | 4 - 5 | 725 | 118 | |
NM_001141978.1 | NP_001135450.1 | 4 - 5 | 524 | 118 | |
NM_028109.4 | NP_082385.3 | 4 - 5 | 712 | 118 | |
GenBank mRNA | AK011311.2 | - | 1 - 2 | 80 | 118 |
AK090101.1 | - | 4 - 5 | 475 | 118 | |
AK153957.1 | - | 4 - 5 | 459 | 118 | |
AK167030.1 | - | 1 - 2 | 177 | 118 | |
BC060619.1 | - | 4 - 5 | 613 | 118 | |
BU148033.1 | - | 4 - 5 | 439 | 118 |