Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Psmc1
Entrez Gene ID
19179
Gene Name
protease (prosome, macropain) 26S subunit, ATPase 1
Gene Aliases
AI325227, P26s4, Rpt2/S4, S4, rpt2
Chromosome Location
Chr.12: 100110202 - 100123364 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008947.3 NP_032973.1 8 - 9 1040 157
GenBank mRNA AK088411.1 - 9 - 10 1070 157
AK133926.1 - 9 - 10 1074 157
AK148906.1 - 9 - 10 1062 157
AK161641.1 - 9 - 10 1089 157
AK161723.1 - 9 - 10 1090 157
AK166783.1 - 9 - 10 1055 157
AK169015.1 - 9 - 10 1053 157
BC003860.1 - 9 - 10 1056 157
U39302.1 - 8 - 9 1041 157

View Full Product Details