Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tmem50a
Entrez Gene ID
71817
Gene Name
transmembrane protein 50A
Gene Aliases
3200001F09Rik, CAM, Smp1
Chromosome Location
Chr.4: 134897849 - 134915030 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027935.2 NP_082211.1 1 - 2 26 64
GenBank mRNA AK014282.1 - 1 - 2 96 64
AK147069.1 - 1 - 2 62 64
AK152630.1 - 1 - 2 110 64
AK152654.1 - 1 - 2 81 64
AK157214.1 - 1 - 2 99 64
AK164989.1 - 1 - 2 92 64
BC094256.1 - 1 - 2 26 64

View Full Product Details