The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177305.4 | NP_796279.2 | 1 - 1 | 3527 | 167 |
GenBank mRNA | AK080743.1 | - | 1 - 1 | 3481 | 167 |