The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026921.4 | NP_081197.1 | 4 - 4 | 956 | 119 |
GenBank mRNA | AK041946.1 | - | 1 - 1 | 155 | 119 |
AK045175.1 | - | 4 - 4 | 940 | 119 | |
AK090000.1 | - | 4 - 4 | 957 | 119 | |
AK153820.1 | - | 4 - 4 | 903 | 119 | |
AK169028.1 | - | 4 - 4 | 927 | 119 | |
AK182282.1 | - | 1 - 1 | 119 | ||
BC018547.1 | - | 4 - 4 | 929 | 119 | |
BC059872.1 | - | 1 - 1 | 2320 | 119 | |
BC085482.1 | - | 4 - 4 | 942 | 119 |