The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007725.2 | NP_031751.1 | 7 - 7 | 1633 | 142 |
GenBank mRNA | AK051845.1 | - | 7 - 7 | 1634 | 142 |
AK208961.1 | - | 1 - 1 | 279 | 142 | |
AK209669.1 | - | 1 - 1 | 279 | 142 | |
AK209768.1 | - | 1 - 1 | 299 | 142 | |
AK218447.1 | - | 1 - 1 | 279 | 142 | |
AK219047.1 | - | 1 - 1 | 142 | ||
BC008181.1 | - | 7 - 7 | 1551 | 142 | |
BC009144.1 | - | 7 - 7 | 1588 | 142 | |
BC018482.1 | - | 7 - 7 | 1603 | 142 | |
CB590846.1 | - | 1 - 1 | 516 | 142 |