The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001136237.1 | NP_001129709.1 | 8 - 9 | 1250 | 66 |
NM_028051.3 | NP_082327.2 | 7 - 8 | 1336 | 66 | |
NM_028092.3 | NP_082368.1 | 7 - 8 | 1212 | 66 | |
GenBank mRNA | AK008448.1 | - | 7 - 8 | 1213 | 66 |
BC028990.1 | - | 8 - 9 | 1211 | 66 |