Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc17a3
Entrez Gene ID
105355
Gene Name
solute carrier family 17 (sodium phosphate), member 3
Gene Aliases
AW261723, Npt4
Chromosome Location
Chr.13: 23836918 - 23860718 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_134069.3 NP_598830.2 1 - 2 227 80
GenBank mRNA AK143896.1 - 1 - 2 199 80
AK165505.1 - 1 - 2 72 80
BC018252.1 - 1 - 2 156 80
BC024459.1 - 1 - 2 183 80
BC034179.1 - 1 - 2 47 80
BC034183.1 - 1 - 2 32 80

View Full Product Details