Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gpld1
Entrez Gene ID
14756
Gene Name
glycosylphosphatidylinositol specific phospholipase D1
Gene Aliases
6330541J12Rik, AW546131
Chromosome Location
Chr.13: 24943152 - 24991936 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008156.2 NP_032182.2 15 - 16 1655 88
GenBank mRNA AK004957.2 - 14 - 15 1359 88
AY081194.1 - 14 - 15 1382 88
AY333960.1 - 14 - 15 1339 88
BC019146.1 - 14 - 15 1368 88

View Full Product Details