Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ncald
Entrez Gene ID
52589
Gene Name
neurocalcin delta
Gene Aliases
AI848120, D030020D09Rik, D15Ertd412e
Chromosome Location
Chr.15: 37366175 - 37792519 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001170866.1 NP_001164337.1 3 - 4 212 64
NM_134094.4 NP_598855.2 3 - 4 402 64
GenBank mRNA AI893360.1 - 3 - 4 208 64
AK048440.1 - 3 - 4 382 64
BY258456.1 - 3 - 4 214 64

View Full Product Details