Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ccdc84
Entrez Gene ID
382073
Gene Name
coiled-coil domain containing 84
Gene Aliases
D630044F24Rik, Gm1114
Chromosome Location
Chr.9: 44410159 - 44418084 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_201372.3 NP_958760.2 2 - 3 301 102
GenBank mRNA AK052757.1 - 2 - 3 316 102
AK162606.1 - 2 - 3 334 102
AK171754.1 - - 308 102
BC055929.1 - 2 - 3 301 102
BC096560.1 - 2 - 3 306 102
BC116877.1 - 2 - 3 315 102
BC116879.1 - 2 - 3 315 102
BC144712.1 - 2 - 3 315 102
BC144713.1 - 2 - 3 315 102

View Full Product Details