Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ubxn7
Entrez Gene ID
224111
Gene Name
UBX domain protein 7
Gene Aliases
A630090P18, AI196514, Ubxd7, mKIAA0794
Chromosome Location
Chr.16: 32332184 - 32393750 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177633.4 NP_808301.3 9 - 10 1269 88
GenBank mRNA AK129218.1 - 5 - 6 867 88
AK147730.1 - 9 - 10 1265 88
BC062904.1 - 8 - 9 1187 88

View Full Product Details