Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cyp2f2
Entrez Gene ID
13107
Gene Name
cytochrome P450, family 2, subfamily f, polypeptide 2
Gene Aliases
Cyp2f
Chromosome Location
Chr.7: 27119955 - 27133660 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007817.2 NP_031843.2 3 - 4 416 75
GenBank mRNA BC011089.1 - 3 - 4 387 75
BC024742.1 - 3 - 4 397 75
BU148924.1 - 3 - 4 380 75
M77497.1 - 3 - 4 396 75

View Full Product Details