Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Idh3a
Entrez Gene ID
67834
Gene Name
isocitrate dehydrogenase 3 (NAD+) alpha
Gene Aliases
1110003P10Rik, 1500012E04Rik, AA407078, AI316514
Chromosome Location
Chr.9: 54586511 - 54604662 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_029573.2 NP_083849.1 9 - 10 872 76
GenBank mRNA AK003393.1 - 6 - 7 765 76
AK010065.1 - 9 - 10 896 76
AK032787.1 - 9 - 10 896 76
AK047951.1 - 9 - 10 894 76
AK117103.1 - 9 - 10 908 76
AK146717.1 - 6 - 7 736 76
AK150618.1 - 9 - 10 903 76
AK151304.1 - 9 - 10 897 76
AK152353.1 - 9 - 10 906 76
AK153459.1 - 9 - 10 897 76
AK158646.1 - 9 - 10 1050 76
AK159051.1 - 9 - 10 969 76
AK164038.1 - 10 - 11 999 76
AK167822.1 - 9 - 10 902 76
AK168034.1 - 9 - 10 894 76
AK168049.1 - 9 - 10 897 76
AK168149.1 - 9 - 10 903 76
AK169152.1 - 9 - 10 870 76
BC034273.1 - 9 - 10 862 76
BC049956.1 - 9 - 10 872 76

View Full Product Details