The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008613.3 | NP_032639.1 | 5 - 6 | 837 | 133 |
GenBank mRNA | AK132772.1 | - | 5 - 6 | 812 | 133 |
BC046624.1 | - | 5 - 6 | 750 | 133 | |
D14849.1 | - | 5 - 6 | 868 | 133 |