The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021050.2 | NP_066388.1 | 12 - 13 | 1810 | 140 |
GenBank mRNA | M60493.1 | - | 12 - 13 | 1811 | 140 |
M69298.1 | - | 12 - 13 | 1809 | 140 |