Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Xpot
Entrez Gene ID
73192
Gene Name
exportin, tRNA (nuclear export receptor for tRNAs)
Gene Aliases
1110004L07Rik, 3110065H13Rik, AI452076, C79645, EXPORTIN-T
Chromosome Location
Chr.10: 121587380 - 121626316 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081056.1 NP_001074525.1 5 - 6 558 71

View Full Product Details