The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001110015.1 | NP_001103485.1 | 3 - 4 | 452 | 75 |
NM_001110016.1 | NP_001103486.1 | 3 - 4 | 452 | 75 | |
NM_144863.4 | NP_659112.2 | 3 - 4 | 452 | 75 | |
GenBank mRNA | AK041763.1 | - | 3 - 4 | 372 | 75 |
AK045396.1 | - | 3 - 4 | 425 | 75 | |
AK077545.1 | - | 3 - 4 | 382 | 75 | |
AK140019.1 | - | 3 - 4 | 377 | 75 | |
AK155965.1 | - | 3 - 4 | 351 | 75 | |
AK156822.1 | - | - | 453 | 75 | |
AK171683.1 | - | 3 - 4 | 390 | 75 | |
AK172054.1 | - | 3 - 4 | 377 | 75 | |
AK172377.1 | - | 3 - 4 | 382 | 75 | |
BC022132.1 | - | 3 - 4 | 424 | 75 | |
BC025809.1 | - | 3 - 4 | 366 | 75 | |
BC094453.1 | - | 3 - 4 | 403 | 75 |