Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mrpl23
Entrez Gene ID
19935
Gene Name
mitochondrial ribosomal protein L23
Gene Aliases
L23mrp, Rpl23, Rpl23l
Chromosome Location
Chr.7: 142533030 - 142540747 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011288.1 NP_035418.1 1 - 2 83 127

View Full Product Details