The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286016.1 | NP_001272945.1 | 9 - 10 | 323 | 77 |
NM_007784.3 | NP_031810.1 | 9 - 10 | 323 | 77 | |
GenBank mRNA | AK052808.1 | - | 9 - 10 | 321 | 77 |
AK085690.1 | - | 9 - 10 | 324 | 77 | |
BC002101.1 | - | 9 - 10 | 292 | 77 | |
BC003859.1 | - | 9 - 10 | 307 | 77 | |
BC007192.1 | - | 9 - 10 | 307 | 77 | |
BC008278.1 | - | 9 - 10 | 307 | 77 | |
BC040246.1 | - | 9 - 10 | 319 | 77 | |
M36780.1 | - | 8 - 9 | 262 | 77 |