Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Clec5a
- Entrez Gene ID
-
23845
- Gene Name
- C-type lectin domain family 5, member a
- Gene Aliases
-
Clecsf5, Ly100, MDL-1, Mdl1
- Chromosome Location
-
Chr.6: 40574898 - 40585805 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK046600.1 |
- |
4 - 5 |
441 |
90 |
View Full Product Details