Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gpld1
Entrez Gene ID
14756
Gene Name
glycosylphosphatidylinositol specific phospholipase D1
Gene Aliases
6330541J12Rik, AW546131
Chromosome Location
Chr.13: 24943152 - 24991936 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008156.2 NP_032182.2 6 - 7 759 80
GenBank mRNA AK004957.2 - 5 - 6 463 80
AY081194.1 - 5 - 6 486 80
AY333960.1 - 5 - 6 443 80
BC019146.1 - 5 - 6 472 80

View Full Product Details