The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001170866.1 | NP_001164337.1 | 2 - 3 | 165 | 82 |
NM_134094.4 | NP_598855.2 | 2 - 3 | 355 | 82 | |
GenBank mRNA | AI893360.1 | - | 2 - 3 | 161 | 82 |
AK048440.1 | - | 2 - 3 | 335 | 82 |