The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011326.2 | NP_035456.1 | 11 - 12 | 1577 | 90 |
GenBank mRNA | AF112187.1 | - | 11 - 12 | 1577 | 90 |
AK143995.1 | - | 10 - 11 | 1270 | 90 | |
BC021338.1 | - | 11 - 12 | 1616 | 90 |