Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Prl8a8
- Entrez Gene ID
-
74188
- Gene Name
- prolactin family 8, subfamily a, member 81
- Gene Aliases
-
1600032B14Rik, AV036230, PLP-Cgamma, Prlpc3
- Chromosome Location
-
Chr.13: 27507032 - 27513228 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AF230923.1 |
- |
2 - 3 |
293 |
177 |
View Full Product Details