Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Mboat1
- Entrez Gene ID
-
218121
- Gene Name
- membrane bound O-acyltransferase domain containing 1
- Gene Aliases
-
9130215M02Rik, BC023845, LPEAT1, Moact1, Oact1
- Chromosome Location
-
Chr.13: 30136449 - 30246694 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK020281.1 |
- |
1 - 2 |
216 |
111 |
View Full Product Details