Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sez6l2
Entrez Gene ID
233878
Gene Name
seizure related 6 homolog like 2
Gene Aliases
AI835913, AW121566, BSRP-A, Psk1
Chromosome Location
Chr.7: 126950043 - 126970610 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001252566.1 NP_001239495.1 12 - 13 2601 85
NM_001252567.1 NP_001239496.1 12 - 13 2421 85
NM_144926.5 NP_659175.1 12 - 13 2601 85
GenBank mRNA AB206789.1 - 12 - 13 2202 85
AK034088.1 - 12 - 13 2573 85
AK165738.1 - 12 - 13 2576 85
BC011475.1 - 12 - 13 2407 85
BC079573.1 - 12 - 13 1990 85
BC096615.1 - 12 - 13 2513 85

View Full Product Details