Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sez6l2
Entrez Gene ID
233878
Gene Name
seizure related 6 homolog like 2
Gene Aliases
AI835913, AW121566, BSRP-A, Psk1
Chromosome Location
Chr.7: 126950043 - 126970610 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001252566.1 NP_001239495.1 15 - 16 3110 70
NM_001252567.1 NP_001239496.1 16 - 17 2969 70
NM_144926.5 NP_659175.1 16 - 17 3149 70
GenBank mRNA AB206789.1 - 16 - 17 2750 70
AK034088.1 - 16 - 17 3121 70
AK165738.1 - 15 - 16 3082 70
BC011475.1 - 16 - 17 2955 70
BC079573.1 - 16 - 17 2538 70
BC096615.1 - 15 - 16 3022 70

View Full Product Details