Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ano9
Entrez Gene ID
71345
Gene Name
anoctamin 9
Gene Aliases
5430425C04Rik, Tmem16j, Tp53i5, Trp53i5
Chromosome Location
Chr.7: 141101218 - 141117806 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178381.3 NP_848468.2 23 - 24 2307 77
GenBank mRNA AK017334.1 - 4 - 5 357 77
BC025477.1 - 22 - 23 2383 77
BC049815.1 - 12 - 13 2413 77
BY717147.1 - 4 - 5 353 77

View Full Product Details