Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Zranb1
- Entrez Gene ID
-
360216
- Gene Name
- zinc finger, RAN-binding domain containing 1
- Gene Aliases
-
9330160G10Rik, D7Wsu87e, Gm1956, Trabid
- Chromosome Location
-
Chr.7: 132931147 - 132986391 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_207302.1 |
NP_997185.1 |
4 - 5 |
1229 |
117 |
View Full Product Details