The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009292.1 | NP_033318.1 | 8 - 9 | 1212 | 71 |
GenBank mRNA | AK006887.1 | - | 9 - 10 | 1553 | 71 |
BC100746.1 | - | 7 - 8 | 960 | 71 | |
BC103590.1 | - | 8 - 9 | 1158 | 71 | |
BC103591.1 | - | 8 - 9 | 1158 | 71 | |
BC103603.1 | - | 8 - 9 | 1155 | 71 | |
Z75287.1 | - | 8 - 9 | 1212 | 71 |