Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Grhl2
Entrez Gene ID
252973
Gene Name
grainyhead-like 2 (Drosophila)
Gene Aliases
0610015A08Rik, BOM, Tcfcp2l3
Chromosome Location
Chr.15: 37233036 - 37363569 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026496.4 NP_080772.2 14 - 15 2050 80
GenBank mRNA AF411213.1 - 14 - 15 1783 80
AK161180.1 - 7 - 8 728 80
BC004783.1 - 5 - 6 456 80
BC041795.1 - 2 - 3 183 80
BC042575.1 - 9 - 10 883 80
BC055035.1 - 14 - 15 1839 80

View Full Product Details